Pentafluorothiophenol ≥98.0%

Leverantör: TCI
Fara

Synonyms: 2,3,4,5,6-Pentafluorothiophenol, Pentafluorophenyl mercaptan, Pentafluorobenzenethiol, Mercaptopentafluorobenzene

P0861-25G P0861-5G
TCIAP0861-25GEA 2450 SEK
TCIAP0861-25G TCIAP0861-5G
Pentafluorothiophenol ≥98.0%
Pentafluorothiophenol
Formel: C₆HF₅S
MW: 200.13 g/mol
Kokpunkt: 143°C
Smältpunkt: –24°C
Flampunkt: 51°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004828
CAS nummer: 771-62-0
EINECS: 212-236-3
UN: 2920
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52f570646f2058a33ef88954e6f19761c9a0e073&vid=52f570646f2058a33ef88954e6f19761c9a0e073&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR