Octadecyl acetate ≥97.0%

Leverantör: TCI

Synonyms: Acetic acid Octadecyl ester, Stearyl acetate, Acetic acid Stearyl ester

A0675-25G A0675-250G
TCIAA0675-25GEA 390 SEK
TCIAA0675-25G TCIAA0675-250G
Octadecyl acetate ≥97.0%
Octadecyl acetate
Formel: C₂₀H₄₀O₂
MW: 312.54 g/mol
Kokpunkt: 344
Smältpunkt: 34.5
Storage Temperature: Kylskåp
MDL Number: MFCD00043649
CAS nummer: 822-23-1
EINECS: 212-493-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c65a9e906b574f7fa883455f7192533bade34586&vid=c65a9e906b574f7fa883455f7192533bade34586&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR