Nonyl acetate ≥99.0%

Leverantör: TCI EUROPE

Synonyms: Acetic acid nonyl ester

A0041-25ML A0041-500ML
TCIAA0041-25MLEA 215 SEK
TCIAA0041-25ML TCIAA0041-500ML
Nonyl acetate ≥99.0%
Nonyl acetate
Formel: C₁₁H₂₂O₂
MW: 186,29 g/mol
Kokpunkt: 228°C
Smältpunkt: –26 °C
Densitet: 0,868 g/cm³ (20 °C)
Flampunkt: 108°C
MDL Number: MFCD00027340
CAS nummer: 143-13-5
EINECS: 205-585-8
Merck Index: 14,06678
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=582bd41a5e6650c9f32e2a652c1081b3f795e678&vid=582bd41a5e6650c9f32e2a652c1081b3f795e678&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR