cis-2-Octene ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
O0098-5ML
O0098-1ML
TCIAO0098-5MLEA
2350
SEK
TCIAO0098-5ML
TCIAO0098-1ML
cis-2-Octene ≥95.0%
cis-2-Octene
Formel:
C₈H₁₆ MW: 112,22 g/mol Kokpunkt: 126°C Smältpunkt: –100,2 °C Densitet: 0,7243 g/cm³ (20 °C) Flampunkt: 21°C |
MDL Number:
MFCD00065146 CAS nummer: 7642-04-8 EINECS: 231-577-9 UN: 3295 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...