Linolsyra ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
L0124-25G L0124-5G
TCIAL0124-25GEA 2425 SEK
TCIAL0124-25G TCIAL0124-5G
Linolsyra ≥97.0% (by GC, titration analysis)
Linolsyra
Formel: C₁₈H₃₂O₂
MW: 280,45 g/mol
Kokpunkt: 177°C
Smältpunkt: –5°C
Densitet: 0,9 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00064241
CAS nummer: 60-33-3
EINECS: 200-470-9
Merck Index: 14,05505
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf291b412effb9342c01769ad8e0b142940020f8&vid=bf291b412effb9342c01769ad8e0b142940020f8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %