Linolsyra ≥85.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
L0053-25ML
L0053-500ML
L0053-100ML
TCIAL0053-25MLEA
605
SEK
TCIAL0053-25ML
TCIAL0053-500ML
TCIAL0053-100ML
Linolsyra ≥85.0%
Linolsyra
Formel:
C₁₈H₃₂O₂ MW: 280,45 g/mol Kokpunkt: 230°C Smältpunkt: -5°C Densitet: 0,9 g/cm³ (20 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00064241 CAS nummer: 60-33-3 EINECS: 200-470-9 Merck Index: 14,05505 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 85.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...