Chlorotriethylsilane ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Triethylchlorosilane
T0589-100G
T0589-25G
T0589-5G
TCIAT0589-100GEA
1700
SEK
TCIAT0589-100G
TCIAT0589-25G
TCIAT0589-5G
Chlorotriethylsilane ≥97.0%
Chlorotriethylsilane
Formel:
C₆H₁₅ClSi MW: 150,72 g/mol Kokpunkt: 149°C Densitet: 0,9 g/cm³ (20 °C) Flampunkt: 30°C Storage Temperature: Omgivande |
MDL Number:
MFCD00000507 CAS nummer: 994-30-9 EINECS: 213-615-6 UN: 2986 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...