8-Bromo-1-octanol ≥90.0%

Leverantör: TCI EUROPE

Synonyms: 8-Bromooctan-1-ol, 1-Bromo-8-hydroxyoctane

B1729-25G B1729-5G
TCIAB1729-25GEA 4525 SEK
TCIAB1729-25G TCIAB1729-5G
8-Bromo-1-octanol ≥90.0%
8-Bromo-1-octanol
Formel: BrH₂C(CH₂)₆CH₂OH
MW: 209,13 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00010388
CAS nummer: 50816-19-8
EINECS: 256-785-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c2e67cb40176b4901eb7d72761ff653dd7c0f8f&vid=6c2e67cb40176b4901eb7d72761ff653dd7c0f8f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR