7-Tridecanone ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Tridecan-7-one, Enanthone, Dihexyl ketone

D1546-25G D1546-5G
TCIAD1546-25GEA 2450 SEK
TCIAD1546-25G TCIAD1546-5G
7-Tridecanone ≥97.0%
7-Tridecanone
Formel: C₁₃H₂₆O
MW: 198,35 g/mol
Kokpunkt: 264 °C (1013 hPa)
Smältpunkt: 30…33 °C
Densitet: 0,825 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00009543
CAS nummer: 462-18-0
EINECS: 207-324-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2712b93e244635b9517e8fbc601f67a5dcbd8276&vid=2712b93e244635b9517e8fbc601f67a5dcbd8276&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Freezing point 27.0 to 31.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR