7-Nitroindole ≥98.0%

Leverantör: TCI EUROPE

N0685-1G N0685-5G
TCIAN0685-1GEA 695 SEK
TCIAN0685-1G TCIAN0685-5G
7-Nitroindole ≥98.0%
7-Nitroindole
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
MDL Number: MFCD00005683
CAS nummer: 6960-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47076bbe44188350bee2432e6b26d89ad131135b&vid=47076bbe44188350bee2432e6b26d89ad131135b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR