7-Methylindole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
M1702-1G
M1702-5G
TCIAM1702-1GEA
630
SEK
TCIAM1702-1G
TCIAM1702-5G
7-Methylindole ≥98.0%
7-Methylindole
Formel:
C₉H₉N MW: 131,18 g/mol Kokpunkt: 266°C Smältpunkt: 83 °C Densitet: 1,0202 g/cm³ (99.6 °C) |
MDL Number:
MFCD00005684 CAS nummer: 933-67-5 EINECS: 213-270-1 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...