6-Nitroindole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Nitro-1H-indole
N0402-1G
N0402-5G
TCIAN0402-1GEA
485
SEK
TCIAN0402-1G
TCIAN0402-5G
6-Nitroindole ≥98.0%
6-Nitroindole
Formel:
C₈H₆N₂O₂ MW: 162,15 g/mol |
MDL Number:
MFCD00051497 CAS nummer: 4769-96-4 EINECS: 225-311-0 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...