5-Hexyn-1-ol ≥95.0%
Leverantör: TCI
Varning
H0687-25ML H0687-5ML
TCIAH0687-25MLEA 2950 SEK
TCIAH0687-25ML TCIAH0687-5ML
5-Hexyn-1-ol ≥95.0%
5-Hexyn-1-ol
Formel: C₆H₁₀O
Kokpunkt: 75°C
Flampunkt: 70°C
Storage Temperature: Kylskåp
MDL Number: MFCD00002980
CAS nummer: 928-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deb2559bf84ebebf3c0d74a23911b56828291fc0&vid=deb2559bf84ebebf3c0d74a23911b56828291fc0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %