6-Chloroindole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Chloro-1H-indole

C1249-1G C1249-5G C1249-25G
TCIAC1249-1GEA 285 SEK
TCIAC1249-1G TCIAC1249-5G TCIAC1249-25G
6-Chloroindole ≥98.0%
6-Chloroindole
Formel: C₈H₆ClN
MW: 151,6 g/mol
Smältpunkt: 90 °C
MDL Number: MFCD00005681
CAS nummer: 17422-33-2
EINECS: 241-449-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a915814fcba38cbc6619cef8d9764aad29ba8738&vid=a915814fcba38cbc6619cef8d9764aad29ba8738&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR