6-Bromoindole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Bromo-1H-indole

B1794-1G B1794-5G
TCIAB1794-1GEA 965 SEK
TCIAB1794-1G TCIAB1794-5G
6-Bromoindole ≥98.0%
6-Bromoindole
Formel: C₈H₆BrN
MW: 196,05 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00238550
CAS nummer: 52415-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a882c056c2c40f66c487ce37fa8d9182f29ca58&vid=2a882c056c2c40f66c487ce37fa8d9182f29ca58&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR