5-Methylindole ≥99.0%
Leverantör: TCI
|
Varning
|
Synonyms:
5-Methyl-1H-indole
M0348-1G
M0348-5G
TCIAM0348-1GEA
365
SEK
TCIAM0348-1G
TCIAM0348-5G
5-Methylindole ≥99.0%
5-Methylindole
Formel:
C₉H₉N Kokpunkt: 100°C Smältpunkt: 60 °C Densitet: 1,0202 g/cm³ (78.3 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00005680 CAS nummer: 614-96-0 EINECS: 210-400-9 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 58.0 to 61.0 deg-C |
Solubility in Methanol | almost transparency |