(±)-5-Methyl-2-hexanol ≥98.0%

Leverantör: TCI EUROPE
Varning

M1666-25G M1666-5G
TCIAM1666-25GEA 815 SEK
TCIAM1666-25G TCIAM1666-5G
(±)-5-Methyl-2-hexanol ≥98.0%
(±)-5-Methyl-2-hexanol
Formel: C₇H₁₆O
MW: 116,2 g/mol
Kokpunkt: 151 °C (1013 hPa)
Densitet: 0,8185 g/cm³ (17.5 °C)
Flampunkt: 46°C
MDL Number: MFCD00004558
CAS nummer: 627-59-8
EINECS: 211-004-9
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3901bbc5a59f41afe1fc1a00a0b35fd2ac4d6585&vid=3901bbc5a59f41afe1fc1a00a0b35fd2ac4d6585&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR