4-Propylpyridine ≥97.0%

Leverantör: TCI EUROPE
Varning

P0669-25ML P0669-5ML
TCIAP0669-25MLEA 1400 SEK
TCIAP0669-25ML TCIAP0669-5ML
4-Propylpyridine ≥97.0%
4-Propylpyridine
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokpunkt: 190°C
Smältpunkt: 4 °C
Densitet: 0,925 g/cm³ (20 °C)
MDL Number: MFCD00014639
CAS nummer: 1122-81-2
EINECS: 214-360-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ee9a412cfd882b2933efd483730e9cb5135aa00&vid=1ee9a412cfd882b2933efd483730e9cb5135aa00&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR