4-Phenylimidazole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Phenyl-1H-imidazole

P0877-25G P0877-5G
TCIAP0877-25GEA 3300 SEK
TCIAP0877-25G TCIAP0877-5G
4-Phenylimidazole ≥98.0%
4-Phenylimidazole
Formel: C₉H₈N₂
MW: 144,18 g/mol
Kokpunkt: 182 - 184 °C (1 -rr)
Smältpunkt: 144…146 °C
MDL Number: MFCD00005197
CAS nummer: 670-95-1
EINECS: 211-580-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5de462cab8f5c46416e6171daf96125534d7af5f&vid=5de462cab8f5c46416e6171daf96125534d7af5f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR