4-Hydroxy-5-decanone ≥97.0%
Leverantör: TCI EUROPE
Synonyms: 6-Hydroxy-5-decanone, Valeroin
V0055-25ML V0055-5ML
TCIAV0055-25MLEA 5775 SEK
TCIAV0055-25ML TCIAV0055-5ML
4-Hydroxy-5-decanone ≥97.0%
4-Hydroxy-5-decanone
Formel: C₁₀H₂₀O₂
MW: 172,27 g/mol
Kokpunkt: 92°C
MDL Number: MFCD00059663
CAS nummer: 6540-98-3
EINECS: 229-455-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71df0bf646675055c686b7741362e469f8d15b9a&vid=71df0bf646675055c686b7741362e469f8d15b9a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %