3-Buten-1-ol ≥98.0%
Leverantör: TCI
Synonyms: But-3-en-1-ol, 4-Hydroxy-1-butene
B0236-10ML B0236-25ML B0236-500ML
TCIAB0236-10MLEA 550 SEK
TCIAB0236-10ML TCIAB0236-25ML TCIAB0236-500ML
3-Buten-1-ol ≥98.0%
3-Buten-1-ol
Formel: C₄H₈O
MDL Number: MFCD00002959
CAS nummer: 627-27-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1fe53d9af2cfbad5a64c087687323f8957b76e4&vid=c1fe53d9af2cfbad5a64c087687323f8957b76e4&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %