4-Fluorobenzoyl chloride ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: p-Fluorobenzoyl chloride

F0213-25G F0213-500G F0213-100G
TCIAF0213-25GEA 285 SEK
TCIAF0213-25G TCIAF0213-500G TCIAF0213-100G
4-Fluorobenzoyl chloride ≥97.0%
4-Fluorobenzoyl chloride
Formel: C₇H₄ClFO
MW: 158,56 g/mol
Kokpunkt: 193°C
Smältpunkt: 9 °C
Densitet: 1,34 g/cm³ (20 °C)
Flampunkt: 82°C
Storage Temperature: Omgivande
MDL Number: MFCD00000684
CAS nummer: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac778407ad243011fce6218904b88ea3773521a7&vid=ac778407ad243011fce6218904b88ea3773521a7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR