4-Acetylpyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl 4-pyridyl ketone
A0113-25G A0113-250G
TCIAA0113-25GEA 445 SEK
TCIAA0113-25G TCIAA0113-250G
4-Acetylpyridine ≥98.0% (by GC, titration analysis)
4-Acetylpyridine
Formel: C₇H₇NO
MW: 121,14 g/mol
Kokpunkt: 214°C
Smältpunkt: 14…16 °C
Densitet: 1,1009 g/cm³ (20 °C)
Flampunkt: 104°C
Storage Temperature: Omgivande
MDL Number: MFCD00006433
CAS nummer: 1122-54-9
EINECS: 214-350-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15b357eb4016faac725e17f27cff5daec9281a4f&vid=15b357eb4016faac725e17f27cff5daec9281a4f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %