4-Cyanobenzaldehyde ≥98.0%

Leverantör: TCI
Varning

C0443-25G C0443-5G
TCIAC0443-25GEA 1100 SEK
TCIAC0443-25G TCIAC0443-5G
4-Cyanobenzaldehyde ≥98.0%
4-Cyanobenzaldehyde
Formel: C₈H₅NO
Kokpunkt: 133°C
Smältpunkt: 100…103 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003376
CAS nummer: 105-07-7
EINECS: 203-267-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3250afff6039a43ab4d6c49711d09e214d38cafb&vid=3250afff6039a43ab4d6c49711d09e214d38cafb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 104.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR