4-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: p-Chlorobenzoyl chloride
C0141-25G C0141-500G C0141-100G
TCIAC0141-25GEA 215 SEK
TCIAC0141-25G TCIAC0141-500G TCIAC0141-100G
4-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Chlorobenzoyl chloride
Formel: C₇H₄Cl₂O
MW: 175.01 g/mol
Kokpunkt: 222°C
Flampunkt: 118°C
Storage Temperature: Omgivande
MDL Number: MFCD00000686
CAS nummer: 122-01-0
EINECS: 204-515-3
UN: 3265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5bdce95f35dfe4f54920f0b7a6c087c5191b7ee&vid=a5bdce95f35dfe4f54920f0b7a6c087c5191b7ee&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %