Methyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl 4-aminobenzoate
A0510-25G A0510-250G
TCIAA0510-25GEA 670 SEK
TCIAA0510-25G TCIAA0510-250G
Methyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)
Methyl-4-aminobenzoate
Formel: C₈H₉NO₂
MW: 151,16 g/mol
Kokpunkt: 299 °C (1013 hPa)
Smältpunkt: 106…107 °C
Densitet: 1,216 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007891
CAS nummer: 619-45-4
EINECS: 210-598-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7506819781a46720650ccc2c40a988aa655d653a&vid=7506819781a46720650ccc2c40a988aa655d653a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 111.0 to 115.0 deg-C
Solubility in Methanol almost transparency