4,6-Decadiyne ≥97.0%
Leverantör: TCI
|
Varning
|
D1726-1ML
D1726-5ML
TCIAD1726-1MLEA
775
SEK
TCIAD1726-1ML
TCIAD1726-5ML
4,6-Decadiyne ≥97.0%
4,6-Decadiyne
Formel:
C₁₀H₁₄ Kokpunkt: 96°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00041645 CAS nummer: 16387-71-6 UN: 3295 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...