2,6-Dihydroxy-9,10-anthraquinone ≥97.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2,6-Dihydroxyanthracene-9,10-dione, Anthraflavic acid, 2,6-Dihydroxyanthraquinone

A1894-1G A1894-5G A1894-25G
TCIAA1894-1GEA 670 SEK
TCIAA1894-1G TCIAA1894-5G TCIAA1894-25G
2,6-Dihydroxy-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
2,6-Dihydroxy-9,10-anthraquinone
Formel: C₁₄H₈O₄
MW: 240.22 g/mol
Smältpunkt: 330°C
MDL Number: MFCD00001228
CAS nummer: 84-60-6
EINECS: 201-544-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58411044f2020be3732c9dca3a7df51768c43b67&vid=58411044f2020be3732c9dca3a7df51768c43b67&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR