3-Phenylpropionic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Phenylpropanoic acid, Hydrocinnamic acid
H0183-25G
H0183-500G
H0183-100G
TCIAH0183-25GEA
215
SEK
TCIAH0183-25G
TCIAH0183-500G
TCIAH0183-100G
3-Phenylpropionic acid ≥98.0% (by GC, titration analysis)
3-Phenylpropionic acid
Formel:
C₉H₁₀O₂ MW: 150,18 g/mol Kokpunkt: 280°C Smältpunkt: 46…47 °C Densitet: 1,071 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00002771 CAS nummer: 501-52-0 EINECS: 207-924-5 Merck Index: 14,04784 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...