(±)-3-Methyl-2-butanol ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
DL-3-Methyl-2-butanol
M0171-25ML
M0171-500ML
TCIAM0171-25MLEA
245
SEK
TCIAM0171-25ML
TCIAM0171-500ML
(±)-3-Methyl-2-butanol ≥98.0%
(±)-3-Methyl-2-butanol
Formel:
C₅H₁₂O Kokpunkt: 112°C Smältpunkt: –117 °C Densitet: 0,82 g/cm³ (20 °C) Flampunkt: 39°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004527 CAS nummer: 598-75-4 EINECS: 209-950-2 UN: 1105 ADR: 3,III Merck Index: 14,06031 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...