3-Fluorobenzoyl chloride 97,0 GC, T
Leverantör: TCI
|
Fara
|
Synonyms:
3-Fluorobenzoylchloride, m-Fluorobenzoyl chloride
F0277-25G
F0277-5G
TCIAF0277-25GEA
1050
SEK
TCIAF0277-25G
TCIAF0277-5G
3-Fluorobenzoyl chloride 97,0 GC, T
3-Fluorobenzoyl chloride
Formel:
C₇H₄ClFO MW: 158.56 g/mol Kokpunkt: 91°C Smältpunkt: -30 Flampunkt: 82°C Storage Temperature: Omgivande |
MDL Number:
MFCD00000670 CAS nummer: 1711-07-5 EINECS: 216-977-3 UN: 3265 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...