3-Etyl-3-pentanol ≥99.0%
Leverantör: TCI EUROPE
E0157-25ML
E0157-100ML
TCIAE0157-25MLEA
1225
SEK
TCIAE0157-25ML
TCIAE0157-100ML
3-Etyl-3-pentanol ≥99.0%
3-Etyl-3-pentanol
Formel:
C₇H₁₆O MW: 116,2 g/mol |
MDL Number:
MFCD00004484 CAS nummer: 597-49-9 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...