5-Chloro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Chloro-6-pyridinol, 5-Chloro-2-pyridinol, 3-Chloro-6-hydroxypyridine

C1245-25G C1245-5G
TCIAC1245-25GEA 750 SEK
TCIAC1245-25G TCIAC1245-5G
5-Chloro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)
5-Chloro-2-hydroxypyridine
Formel: C₅H₄ClNO
MW: 129,55 g/mol
Smältpunkt: 163 °C
MDL Number: MFCD00006275
CAS nummer: 4214-79-3
EINECS: 224-146-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f91c49a2d7639b07a01806ea68b9c74040c6bba9&vid=0ee1290f9688db7a6ecb3d0f8bd2b6b20feeae44&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 163.0 to 166.0 deg-C
Solubility in hot Water within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR