3-Chloro-2-fluorobenzaldehyde ≥95.0%
Leverantör: TCI
|
Varning
|
C2127-25G
C2127-5G
TCIAC2127-25GEA
3000
SEK
TCIAC2127-25G
TCIAC2127-5G
3-Chloro-2-fluorobenzaldehyde ≥95.0%
3-Chloro-2-fluorobenzaldehyde
Formel:
C₇H₄ClFO MW: 158.56 g/mol Flampunkt: 86°C |
MDL Number:
MFCD01631571 CAS nummer: 85070-48-0 |
|
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...