3-Bromopropionic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 3-Bromopropanoic acid

B0645-25G B0645-500G B0645-100G
TCIAB0645-25GEA 215 SEK
TCIAB0645-25G TCIAB0645-500G TCIAB0645-100G
3-Bromopropionic acid ≥98.0% (by GC, titration analysis)
3-Bromopropionic acid
Formel: BrH₂CCH₂COOH
MW: 152,98 g/mol
Kokpunkt: 142°C
Smältpunkt: 60 °C
Densitet: 1,48 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002763
CAS nummer: 590-92-1
EINECS: 209-694-1
UN: 3261
ADR: 8,III
Merck Index: 14,01433
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd19f12265976b3c7368e70449bf62249fcbebba&vid=fd19f12265976b3c7368e70449bf62249fcbebba&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 61.0 to 64.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR