3-Bromo-4-methylpyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Bromo-4-picoline
B2831-1G
B2831-25G
B2831-5G
TCIAB2831-1GEA
215
SEK
TCIAB2831-1G
TCIAB2831-25G
TCIAB2831-5G
3-Bromo-4-methylpyridine ≥98.0% (by GC, titration analysis)
3-Bromo-4-methylpyridine
Formel:
C₆H₆BrN MW: 172,02 g/mol Kokpunkt: 76°C Densitet: 1,549 g/cm³ (20 °C) Flampunkt: 79°C |
MDL Number:
MFCD00082592 CAS nummer: 3430-22-6 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...