3-Bromo-1-propanol ≥93.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane

B0641-25G B0641-250G
TCIAB0641-25GEA 1275 SEK
TCIAB0641-25G TCIAB0641-250G
3-Bromo-1-propanol ≥93.0%
3-Bromo-1-propanol
Formel: BrH₂CCH₂CH₂OH
MW: 138,99 g/mol
Kokpunkt: 140°C
Densitet: 1,6 g/cm³ (20 °C)
Flampunkt: 102°C
Storage Temperature: Kylskåp
MDL Number: MFCD00002942
CAS nummer: 627-18-9
EINECS: 210-986-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7de71ecffe92a2b11065429627b4248fc1fa607d&vid=7de71ecffe92a2b11065429627b4248fc1fa607d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR