5-Amino-2-bromopyridine ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Bromopyridin-3-amine, 2-Bromopyridin-5-amine, 3-Amino-6-bromopyridine, 2-Bromo-5-pyridylamine

A1890-1G A1890-5G
TCIAA1890-1GEA 295 SEK
TCIAA1890-1G TCIAA1890-5G
5-Amino-2-bromopyridine ≥97.0%
5-Amino-2-bromopyridine
Formel: C₅H₅BrN₂
MW: 173,01 g/mol
Kokpunkt: 180°C
MDL Number: MFCD01646116
CAS nummer: 13534-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f7e9d4a3e7f8972bd88fd1b4e77c959299482f&vid=e9f7e9d4a3e7f8972bd88fd1b4e77c959299482f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 97.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR