3,4,5-Trikloropyridin ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

T2714-25G T2714-5G
TCIAT2714-25GEA 3575 SEK
TCIAT2714-25G TCIAT2714-5G
3,4,5-Trikloropyridin ≥98.0% (by GC, titration analysis)
3,4,5-Trikloropyridin
Formel: C₅H₂Cl₃N
MW: 182,44 g/mol
Kokpunkt: 215°C
MDL Number: MFCD00051685
CAS nummer: 33216-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82b462ab5d0cd6ba5a79db9bede902922f1e1f5f&vid=82b462ab5d0cd6ba5a79db9bede902922f1e1f5f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point gt. 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR