2-Ethyl-1-hexene ≥95.0%

Leverantör: TCI
Fara

E0124-10ML E0124-25ML
TCIAE0124-10MLEA 1425 SEK
TCIAE0124-10ML TCIAE0124-25ML
2-Ethyl-1-hexene ≥95.0%
2-Ethyl-1-hexene
Formel: C₈H₁₆
MW: 112.22 g/mol
Kokpunkt: 120°C
Smältpunkt: -109
MDL Number: MFCD00027242
CAS nummer: 1632-16-2
EINECS: 216-636-9
UN: 3295
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1c95223474fde26eeb93b931b4db38f6390683d&vid=b1c95223474fde26eeb93b931b4db38f6390683d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR