2-Ethoxynaphthalene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Ethyl 2-naphthyl ether

N0431-25G N0431-500G N0431-100G
TCIAN0431-25GEA 215 SEK
TCIAN0431-25G TCIAN0431-500G TCIAN0431-100G
2-Ethoxynaphthalene ≥98.0%
2-Ethoxynaphthalene
Formel: C₁₂H₁₂O
MW: 172,23 g/mol
Kokpunkt: 282°C
Smältpunkt: 37 °C
Densitet: 1,064 g/cm³ (20 °C)
MDL Number: MFCD00016808
CAS nummer: 93-18-5
EINECS: 202-226-7
Merck Index: 14,03752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7fdc19a1512307806610b35b9f92130433af67e&vid=f7fdc19a1512307806610b35b9f92130433af67e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 35.0 to 38.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR