2-Chloroheptane ≥97.0%

Leverantör: TCI
Varning

Synonyms: 2-Heptyl chloride

C0852-25ML C0852-5ML
TCIAC0852-25MLEA 3075 SEK
TCIAC0852-25ML TCIAC0852-5ML
2-Chloroheptane ≥97.0%
2-Chloroheptane
Formel: C₇H₁₅Cl
Kokpunkt: 146°C
MDL Number: MFCD00039353
CAS nummer: 1001-89-4
EINECS: 213-683-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b4c3a7c05fa8148ff5a2ce91e2ff6d44dae568f&vid=8b4c3a7c05fa8148ff5a2ce91e2ff6d44dae568f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR