Etanolamin ≥99.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: 2-Aminoetanol
A0297-500G A0297-25G
TCIAA0297-500GEA 270 SEK
TCIAA0297-500G TCIAA0297-25G
Etanolamin ≥99.0% (by GC, titration analysis)
Etanolamin
Formel: NH₂CH₂CH₂OH
MW: 61,08 g/mol
Kokpunkt: 170°C
Smältpunkt: 10,5 °C
Densitet: 1,0164 g/cm³ (20 °C)
Flampunkt: 93°C
Storage Temperature: Omgivande
MDL Number: MFCD00008183
CAS nummer: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8,III
Merck Index: 14,03727
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0732f018521b8da49b3ab21487c9f6d1db64330e&vid=0732f018521b8da49b3ab21487c9f6d1db64330e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %