2,4-Hexadiyne ≥98.0%
Leverantör: TCI EUROPE
Varning
H0502-1G H0502-5G
TCIAH0502-1GEA 1500 SEK
TCIAH0502-1G TCIAH0502-5G
2,4-Hexadiyne ≥98.0%
2,4-Hexadiyne
Formel: C₆H₆
MW: 78,11 g/mol
Kokpunkt: 129°C
Smältpunkt: –67,8 °C
Flampunkt: 35°C
MDL Number: MFCD00041617
CAS nummer: 2809-69-0
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ad11d84e512e7c7f94f375922db6070ad5efaf0&vid=8ad11d84e512e7c7f94f375922db6070ad5efaf0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 67.0 deg-C
Solubility in Methanol very faint turbidity