2,4,6-Trifluorobenzaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning

T2320-1G T2320-5G
TCIAT2320-1GEA 270 SEK
TCIAT2320-1G TCIAT2320-5G
2,4,6-Trifluorobenzaldehyde ≥98.0%
2,4,6-Trifluorobenzaldehyde
Formel: C₇H₃F₃O
MW: 160,1 g/mol
MDL Number: MFCD00061197
CAS nummer: 58551-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86d072535cd9e9c499eac950d274b33bedb3ac51&vid=86d072535cd9e9c499eac950d274b33bedb3ac51&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR