2,3-Butanedithiol ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
2,3-Dimercaptobutane
B2888-25G
B2888-5G
TCIAB2888-25GEA
1950
SEK
TCIAB2888-25G
TCIAB2888-5G
2,3-Butanedithiol ≥98.0%
2,3-Butanedithiol
Formel:
C₄H₁₀S₂ MW: 122.26 g/mol Kokpunkt: 87°C Flampunkt: 52°C |
MDL Number:
MFCD00010024 CAS nummer: 4532-64-3 EINECS: 224-870-8 UN: 3336 ADR: 3 III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...