12-Bromo-1-dodecanol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 12-Bromododecan-1-ol

B1731-25G B1731-5G
TCIAB1731-25GEA 2525 SEK
TCIAB1731-25G TCIAB1731-5G
12-Bromo-1-dodecanol ≥98.0%
12-Bromo-1-dodecanol
Formel: HO(CH₂)₁₁CH₂Br
MW: 265,23 g/mol
Kokpunkt: 156°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004754
CAS nummer: 3344-77-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19791638e9dcad6b238ac3eb96934f12f2167cb8&vid=19791638e9dcad6b238ac3eb96934f12f2167cb8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 31.0 to 35.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR