11-Bromo-1-undecanol ≥97.0%
Leverantör: TCI EUROPE
Synonyms: 11-Bromoundecan-1-ol, 11-Bromoundecanol
B1477-25G B1477-5G
TCIAB1477-25GEA 870 SEK
TCIAB1477-25G TCIAB1477-5G
11-Bromo-1-undecanol ≥97.0%
11-Bromo-1-undecanol
Formel: HO(CH₂)₁₀CH₂Br
MW: 251,21 g/mol
Kokpunkt: 170°C
Smältpunkt: 50 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00004752
CAS nummer: 1611-56-9
EINECS: 216-554-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04b90aa29e16edaedb92af60ec69d50867f4d45b&vid=04b90aa29e16edaedb92af60ec69d50867f4d45b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 45.0 to 49.0 deg-C
Solubility in Methanol almost transparency