4-Toluenethiol ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Mercapto-4-methylbenzene, 4-Methylbenzene-1-thiol, 4-Thiocresol, 4-Methylbenzenethiol, p-Toluenethiol, p-Thiocresol

T0290-25G T0290-500G
TCIAT0290-25GEA 350 SEK
TCIAT0290-25G TCIAT0290-500G
4-Toluenethiol ≥97.0% (by GC, titration analysis)
4-Toluenethiol
Formel: C₇H₈S
MW: 124,21 g/mol
Kokpunkt: 196°C
Smältpunkt: 39…42 °C
Densitet: 1,022 g/cm³ (51 °C)
Flampunkt: 70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004851
CAS nummer: 106-45-6
EINECS: 203-399-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,09325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=917f0754391973827e482aee051592aced65dc9a&vid=917f0754391973827e482aee051592aced65dc9a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %
Melting point 42.0 to 45.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR