3-Toluenethiol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: m-Thiocresol, 3-Methylbenzene-1-thiol, 3-Thiocresol, 3-Methylbenzenethiol, m-Toluenethiol, 1-Mercapto-3-methylbenzene, 3-Methylthiophenol

T0289-25G T0289-5G
TCIAT0289-25GEA 1925 SEK
TCIAT0289-25G TCIAT0289-5G
3-Toluenethiol ≥98.0% (by GC, titration analysis)
3-Toluenethiol
Formel: C₇H₈S
MW: 124,21 g/mol
Kokpunkt: 195°C
Densitet: 1,06251 g/cm³ (0 °C)
Flampunkt: 73°C
MDL Number: MFCD00004843
CAS nummer: 108-40-7
EINECS: 203-578-4
UN: 2810
ADR: 6.1,III
Merck Index: 14,09325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c40b40bf63377e12a257725a7d4cc50ff312bf4&vid=2c40b40bf63377e12a257725a7d4cc50ff312bf4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR